COM448687405 Combi-Blocks, Inc.

methyl 4-(3-chloro-5-fluorophenyl)-3-fluorobenzoate

  • MDL number MFCD27578914
  • Cas Number : 1621256-29-8
  • Molecular Weight : 282.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2618
Availability details for
Quantity
  • Availability
  • Add to Cart