COM448687411 Combi-Blocks, Inc.

[2-ethoxy-5-(methoxycarbonyl)phenyl]boronic acid

  • MDL number MFCD27578919
  • Molecular Weight : 224.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2629
Availability details for
Quantity
  • Availability
  • Add to Cart