COM448687414 Combi-Blocks, Inc.

4-Methyl-biphenyl-2-carboxylic acid methyl ester

  • MDL number MFCD23134172
  • Cas Number : 152620-33-2
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2632

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2632
Availability details for
Quantity
  • Availability
  • Add to Cart