COM448687415 Combi-Blocks, Inc.

methyl 4-(6-chloro-2-methylpyridin-3-yl)benzoate

  • MDL number MFCD27578920
  • Cas Number : 1820685-76-4
  • Molecular Weight : 261.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2633
Availability details for
Quantity
  • Availability
  • Add to Cart