COM448687427 Combi-Blocks, Inc.

methyl 2-(3-hydroxyphenyl)benzoate

  • MDL number MFCD27578929
  • Cas Number : 1251836-88-0
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2650
Availability details for
Quantity
  • Availability
  • Add to Cart