COM448687437 Combi-Blocks, Inc.

methyl 2-(4-{[(tert-butoxy)carbonyl]amino}phenyl)benzoate

  • MDL number MFCD27578934
  • Cas Number : 1820687-23-7
  • Molecular Weight : 327.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2664
Availability details for
Quantity
  • Availability
  • Add to Cart