COM448687439 Combi-Blocks, Inc.

methyl 2-hydroxy-5-phenylpyridine-3-carboxylate

  • MDL number MFCD27578936
  • Cas Number : 76876-86-3
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2667
Availability details for
Quantity
  • Availability
  • Add to Cart