COM448687443 Combi-Blocks, Inc.

methyl (2R)-2-{[(benzyloxy)carbonyl]amino}-3-[4-(benzyloxy)phenyl]propanoate

  • MDL number MFCD27578939
  • Cas Number : 174471-32-0
  • Molecular Weight : 419.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2673
Availability details for
Quantity
  • Availability
  • Add to Cart