COM448687444 Combi-Blocks, Inc.

2,6-Dichloro-3-fluorobenzoic acid methyl ester

  • MDL number MFCD12406864
  • Cas Number : 1214353-49-7
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2674
Availability details for
Quantity
  • Availability
  • Add to Cart