COM448687445 Combi-Blocks, Inc.

methyl 2-fluoro-4-(3-fluorophenyl)benzoate

  • MDL number MFCD27578940
  • Cas Number : 1820711-88-3
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2675
Availability details for
Quantity
  • Availability
  • Add to Cart