COM448687448 Combi-Blocks, Inc.

Methyl 3-(4-bromophenyl)-3-cyanopropanoate

  • MDL number MFCD19362681
  • Cas Number : 1267930-49-3
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2681
Availability details for
Quantity
  • Availability
  • Add to Cart