COM448687449 Combi-Blocks, Inc.

Methyl 6-chlorohexanoate

  • MDL number MFCD09751140
  • Cas Number : 14273-89-3
  • Molecular Weight : 164.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2682
Availability details for
Quantity
  • Availability
  • Add to Cart