COM448687450 Combi-Blocks, Inc.

methyl 2-fluoro-4-[3-methoxy-5-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578942
  • Cas Number : 1820603-46-0
  • Molecular Weight : 318.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2683
Availability details for
Quantity
  • Availability
  • Add to Cart