COM448687459 Combi-Blocks, Inc.

methyl 4-(5-methoxypyridin-3-yl)benzoate

  • MDL number MFCD27578945
  • Cas Number : 1820612-90-5
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2699
Availability details for
Quantity
  • Availability
  • Add to Cart