COM448687461 Combi-Blocks, Inc.

methyl 2-fluoro-4-(6-methoxypyridin-3-yl)benzoate

  • MDL number MFCD27578947
  • Cas Number : 1820704-98-0
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2701
Availability details for
Quantity
  • Availability
  • Add to Cart