COM448687462 Combi-Blocks, Inc.

1-methyl 4-prop-2-en-1-yl (2R)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578948
  • Cas Number : 368443-50-9
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2702

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2702
Availability details for
Quantity
  • Availability
  • Add to Cart