COM448687463 Combi-Blocks, Inc.

methyl 4-fluoro-3-(4-fluoro-3-methylphenyl)benzoate

  • MDL number MFCD27578949
  • Cas Number : 1820740-95-1
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2703
Availability details for
Quantity
  • Availability
  • Add to Cart