COM448687464 Combi-Blocks, Inc.

hydrate methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoate hydrochloride

  • MDL number MFCD27578950
  • Molecular Weight : 342.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2704
Availability details for
Quantity
  • Availability
  • Add to Cart