COM448687466 Combi-Blocks, Inc.

methyl (2S)-1-[(2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoyl]pyrrolidine-2-carboxylate

  • MDL number MFCD01632085
  • Cas Number : 51827-14-6
  • Molecular Weight : 362.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2706
Availability details for
Quantity
  • Availability
  • Add to Cart