COM448687468 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoate

  • MDL number MFCD27578953
  • Cas Number : 1820581-49-4
  • Molecular Weight : 454.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2708
Availability details for
Quantity
  • Availability
  • Add to Cart