COM448687469 Combi-Blocks, Inc.

methyl (2R)-3-[4-(tert-butoxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578954
  • Cas Number : 1820576-23-5
  • Molecular Weight : 473.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2709
Availability details for
Quantity
  • Availability
  • Add to Cart