COM448687470 Combi-Blocks, Inc.

Methyl (2S)-5-[(1E)-(Cbz-amino),(Cbz-imino)methyl]amino-2-Boc-aminopentanoate

  • MDL number MFCD27578955
  • Cas Number : 899442-96-7
  • Molecular Weight : 556.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2710
Availability details for
Quantity
  • Availability
  • Add to Cart