COM448687472 Combi-Blocks, Inc.

dicyclohexylamine methyl (2S)-3-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578957
  • Molecular Weight : 533.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2712

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2712
Availability details for
Quantity
  • Availability
  • Add to Cart