COM448687473 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-{3-[(4-methoxy-2,3,6-trimethylbenzene)sulfonyl]carbamimidamido}pentanoate

  • MDL number MFCD27578958
  • Cas Number : 1820575-89-0
  • Molecular Weight : 500.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2713
Availability details for
Quantity
  • Availability
  • Add to Cart