COM448687475 Combi-Blocks, Inc.

6-Ethoxy-1,3-benzoxazol-2-amine

  • MDL number MFCD20693880
  • Cas Number : 13895-09-5
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3000
Availability details for
Quantity
  • Availability
  • Add to Cart