COM448687480 Combi-Blocks, Inc.

5-Bromo-6-fluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD24203823
  • Cas Number : 1545756-38-4
  • Molecular Weight : 231.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3005
Availability details for
Quantity
  • Availability
  • Add to Cart