COM448687482 Combi-Blocks, Inc.

5-Bromo-7-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD18294017
  • Cas Number : 1268110-58-2
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3007
Availability details for
Quantity
  • Availability
  • Add to Cart