COM448687483 Combi-Blocks, Inc.

4-Methoxy-1,3-benzoxazol-2-amine

  • MDL number MFCD20706975
  • Cas Number : 727415-76-1
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3008
Availability details for
Quantity
  • Availability
  • Add to Cart