COM448687492 Combi-Blocks, Inc.

2-Amino-benzooxazole-5-carboxylic acid ethyl ester

  • MDL number MFCD22193343
  • Cas Number : 56350-62-0
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3017
Availability details for
Quantity
  • Availability
  • Add to Cart