COM448687497 Combi-Blocks, Inc.

6-Bromo-5-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD19376459
  • Cas Number : 1267996-62-2
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3022

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3022
Availability details for
Quantity
  • Availability
  • Add to Cart