COM448687500 Combi-Blocks, Inc.

1,3-Benzoxazole-2,6-diamine

  • MDL number MFCD22193573
  • Cas Number : 1503293-58-0
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3025
Availability details for
Quantity
  • Availability
  • Add to Cart