COM448687501 Combi-Blocks, Inc.

7-Bromo-5-chloro-1,3-benzoxazol-2-amine

  • MDL number MFCD16742786
  • Cas Number : 1247830-45-0
  • Molecular Weight : 247.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3026
Availability details for
Quantity
  • Availability
  • Add to Cart