COM448687502 Combi-Blocks, Inc.

4-Chlorobenzo[d]oxazol-2-amine

  • MDL number MFCD01664217
  • Cas Number : 64037-10-1
  • Molecular Weight : 168.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3027
Availability details for
Quantity
  • Availability
  • Add to Cart