COM448687503 Combi-Blocks, Inc.

6-Chloro-5-fluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD19374814
  • Cas Number : 1268032-07-0
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3028
Availability details for
Quantity
  • Availability
  • Add to Cart