COM448687507 Combi-Blocks, Inc.

6-Nitro-1,3-benzoxazol-2-amine

  • MDL number MFCD11844837
  • Cas Number : 6458-17-9
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3032
Availability details for
Quantity
  • Availability
  • Add to Cart