COM448687509 Combi-Blocks, Inc.

5,6-Dichloro-1,3-benzoxazol-2-amine

  • MDL number MFCD01664141
  • Cas Number : 64037-12-3
  • Molecular Weight : 203.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3034
Availability details for
Quantity
  • Availability
  • Add to Cart