COM448687521 Combi-Blocks, Inc.

5,7-Difluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD16742779
  • Cas Number : 1247508-62-8
  • Molecular Weight : 170.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3046
Availability details for
Quantity
  • Availability
  • Add to Cart