COM448687524 Combi-Blocks, Inc.

5-Methanesulfonyl-1,3-benzoxazol-2-amine

  • MDL number MFCD24105804
  • Cas Number : 1507862-91-0
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3049
Availability details for
Quantity
  • Availability
  • Add to Cart