COM448687539 Combi-Blocks, Inc.

methyl 2-amino-5-bromo-1,3-benzoxazole-7-carboxylate

  • MDL number MFCD27936870
  • Cas Number : 1820704-53-7
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3064
Availability details for
Quantity
  • Availability
  • Add to Cart