COM448687541 Combi-Blocks, Inc.

2-amino-1,3-benzoxazole-4-carbonitrile

  • MDL number MFCD27936872
  • Cas Number : 1806318-91-1
  • Molecular Weight : 159.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3066
Availability details for
Quantity
  • Availability
  • Add to Cart