COM448687542 Combi-Blocks, Inc.

6-(Trifluoromethyl)-1,3-benzoxazol-2-amine

  • MDL number MFCD27936873
  • Cas Number : 747353-64-6
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3067
Availability details for
Quantity
  • Availability
  • Add to Cart