COM448687548 Combi-Blocks, Inc.

2-amino-7-bromo-1,3-benzoxazole-5-carboxylic acid

  • MDL number MFCD27936878
  • Cas Number : 1820608-61-4
  • Molecular Weight : 257.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3073
Availability details for
Quantity
  • Availability
  • Add to Cart