COM448687551 Combi-Blocks, Inc.

5,7-dibromo-6-fluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD27936880
  • Cas Number : 1820612-64-3
  • Molecular Weight : 309.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3076
Availability details for
Quantity
  • Availability
  • Add to Cart