COM448687552 Combi-Blocks, Inc.

2-amino-7-methoxy-1,3-benzoxazole-5-carbaldehyde

  • MDL number MFCD27936881
  • Cas Number : 1820607-78-0
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3077
Availability details for
Quantity
  • Availability
  • Add to Cart