COM448687562 Combi-Blocks, Inc.

2-amino-6-fluoro-1,3-benzoxazole-7-carbonitrile

  • MDL number MFCD27936887
  • Cas Number : 1820648-99-4
  • Molecular Weight : 177.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3087
Availability details for
Quantity
  • Availability
  • Add to Cart