COM448687565 Combi-Blocks, Inc.

5-methyl-1,3-benzoxazole-2,6-diamine

  • MDL number MFCD27936890
  • Cas Number : 1820683-80-4
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3090
Availability details for
Quantity
  • Availability
  • Add to Cart