COM448687569 Combi-Blocks, Inc.

2-amino-7-bromo-1,3-benzoxazole-5-carboxamide

  • MDL number MFCD27936893
  • Cas Number : 1820707-28-5
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3094
Availability details for
Quantity
  • Availability
  • Add to Cart