COM448687570 Combi-Blocks, Inc.

5-benzoyl-1,3-benzoxazol-2-amine

  • MDL number MFCD27936894
  • Cas Number : 1686143-09-8
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3095

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3095
Availability details for
Quantity
  • Availability
  • Add to Cart