COM448687571 Combi-Blocks, Inc.

5-methyl-1,3-benzoxazole-2,4-diamine

  • MDL number MFCD27936895
  • Cas Number : 1820683-86-0
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3096
Availability details for
Quantity
  • Availability
  • Add to Cart