COM448687572 Combi-Blocks, Inc.

2-amino-1,3-benzoxazole-5-carboxamide

  • MDL number MFCD27936896
  • Cas Number : 1806532-37-5
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3097
Availability details for
Quantity
  • Availability
  • Add to Cart